Skip to Content
Merck
All Photos(1)

Key Documents

Z514500

Safeskin® purple nitrile gloves

size XL

Synonym(s):

disposable glove

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35
Pricing and availability is not currently available.

material

nitrile
purple

size

XL

sterility

non-sterile

feature

disposable
powder-free
latex free

packaging

pack of 100 ea (boxes of 100)
pk of 100 ea

manufacturer/tradename

Kimberly Clark 55084

availability

not available in EU

L

9 1/2 in.

thickness

4.7 mil , palm
5.9 mil , fingertips

color

purple

Looking for similar products? Visit Product Comparison Guide

General description

These unique purple gloves combine the durability of nitrile with superb comfort and fit. Powder free design reduces the risk of powder-related interference during lab work

Legal Information

Safeskin is a registered trademark of Kimberly-Clark Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service