Key Documents
Z272620
Flow control pinch valve
for tubing O.D. 7/16 - 1/2in.
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
material
Delrin® body
General description
Metered scale on valve body adjusts fluid from zero to full flow without fluid contact with valve. Calibration is accurate and repeatable. Valve mounts easily on existing soft wall tube lines without fittings or cutting.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)