Skip to Content
Merck
All Photos(1)

Documents

Z137197

Supelco

Ace chromatography column

I.D. × L 50 mm × 61 cm, With fritted disc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

grade

for chromatography

manufacturer/tradename

Ace Glass 590549

technique(s)

LPLC: suitable

I.D. × L

50 mm × 61 cm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Has #15 Ace-Thred at top and 2 mm bore, solid-PTFE stopcock-plug (tapered 1:5) at bottom. Supplied with nylon and FETFE® O-ring for connecting 12.5 to 14 mm O.D. tube to column, and with tubing connector.

Note:
When using tubing connector, O-ring supplied with bushing is NOT necessary. Fritted disc has 70 to 100 μm porosity, sealed into bottom of column.

Legal Information

FETFE is a registered trademark of Ace Glass, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service