Skip to Content
Merck
All Photos(1)

Key Documents

W521108

Sigma-Aldrich

Bran absolute

FG

Synonym(s):

Bran

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164502

biological source

Triticum estivum

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

origin

France origin

refractive index

n20/D 1.476 (lit.)

density

0.945 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Preparation Note

Extraction method: solvent extraction

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service