Skip to Content
Merck
All Photos(1)

Key Documents

W214520

Sigma-Aldrich

Benzyl formate

natural, 97%, FG

Synonym(s):

Benzyl methanoate, Formic acid benzyl ester, NSC 8049

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH2C6H5
CAS Number:
Molecular Weight:
136.15
FEMA Number:
2145
Beilstein:
150-84-5
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.435
NACRES:
NA.21
Organoleptic:
fruity; pineapple; sweet
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

97%

refractive index

n20/D 1.511 (lit.)

bp

203 °C (lit.)

density

1.088 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; pineapple; sweet

SMILES string

O=COCC1=CC=CC=C1

InChI

1S/C8H8O2/c9-7-10-6-8-4-2-1-3-5-8/h1-5,7H,6H2

InChI key

UYWQUFXKFGHYNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service