Key Documents
TEM-76043-22
Sigma-Aldrich
Cryo-SiN TEM Windows
20 nm Thick, window size 500 μm , Single
About This Item
UNSPSC Code:
41111761
window size
500 μm , Single
General description
Silicon Nitride 20 nm thick TEM Windows - Single 500 micron window. Ideal substrate for Cryo-Microscopy. Avoid substrate degradation. Plasma cleanable to reduce contaminants. Large 500 micron windows provid 50x50 micron viewing from any orientation at 70 degreee of tilt for tomography.
Choose from one of the most recent versions:
Certificates of Analysis (COA)