SYNM250005
Synthware™ all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 5, Hose Connections: Front-left-right, Rear-left-right
All Photos(1)
About This Item
Recommended Products
feature
number of ports 5
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M250005
Related Categories
General description
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left-right and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Legal Information
Synthware is a trademark of Kemtech America Inc.