SYNA351424
Synthware™ distillation adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 24/40, side joint: 24/40, side arm angle: 75 deg
Synonym(s):
Synthware A351424, adapter, sidearm adapter
All Photos(1)
About This Item
Recommended Products
Joint
bottom joint: ST/NS24/40
side joint: 24/40
joint: ST/NS top14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A351424
Related Categories
General description
For use as a component in atmospheric or vacuum distillations.
Legal Information
Synthware is a trademark of Kemtech America Inc.