All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H7NS
CAS Number:
Molecular Weight:
101.17
Beilstein:
104274
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.520 (lit.)
bp
144.0-145.0 °C (lit.)
mp
-101.0 °C (lit.)
density
1.067 g/mL at 25 °C (lit.)
SMILES string
CC1=NCCS1
InChI
1S/C4H7NS/c1-4-5-2-3-6-4/h2-3H2,1H3
InChI key
JUIQOABNSLTJSW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Meyers reagent for aldehyde synthesis.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
98.6 °F - closed cup
Flash Point(C)
37 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B Schaal et al.
Comptes rendus de l'Academie des sciences. Serie III, Sciences de la vie, 313(7), 319-325 (1991-01-01)
Intranasal injections of a fragrant solution of 2-methyl-2-thiazoline elicited significant heart rate decelerations in late pregnancy fetal sheep, while the injection of a control fluid (isotonic saline) had no effect. This result indicates that the ovine fetuses are able to
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (10)(10), 55-56 (1980-10-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service