Skip to Content
Merck
All Photos(1)

Key Documents

M3000

Sigma-Aldrich

(−)-Menthyloxyacetic acid

98%

Synonym(s):

(−)-Menthyl carboxymethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O3
CAS Number:
Molecular Weight:
214.30
Beilstein:
2444474
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]25/D −92.5°, c = 4 in methanol

optical purity

ee: 97% (GLC)

refractive index

n20/D 1.4672 (lit.)

bp

163-164 °C/10 mmHg (lit.)

mp

52-55 °C (lit.)

density

1.01 g/mL at 20 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OCC(O)=O

InChI

1S/C12H22O3/c1-8(2)10-5-4-9(3)6-11(10)15-7-12(13)14/h8-11H,4-7H2,1-3H3,(H,13,14)/t9-,10+,11-/m1/s1

InChI key

CILPHQCEVYJUDN-OUAUKWLOSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service