Skip to Content
Merck
All Photos(2)

Key Documents

I18202

Sigma-Aldrich

2-Isonitrosoacetophenone

97%

Synonym(s):

ω-(Hydroxyimino)acetophenone, ω-Isonitrosoacetophenone, Phenylglyoxaldoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH=NOH
CAS Number:
Molecular Weight:
149.15
Beilstein:
2041691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

126-128 °C (lit.)

storage temp.

2-8°C

SMILES string

O\N=C\C(=O)c1ccccc1

InChI

1S/C8H7NO2/c10-8(6-9-11)7-4-2-1-3-5-7/h1-6,11H/b9-6+

InChI key

MLNKXLRYCLKJSS-RMKNXTFCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I A Dubery et al.
Phytochemistry, 50(6), 983-989 (1999-07-01)
The time- and dose-dependent occurrence of 4-(3-methyl-2-butenoxy)isonitrosoacetophenone, a gamma-irradiation-induced stress metabolite was investigated. The chemical synthesis of the compound is reported. The compound exhibits antifungal activity, as well as antioxidant activity, as indicated by its ability to scavenge reactive oxygen
C Meredith et al.
Journal of neurochemistry, 51(4), 1097-1101 (1988-10-01)
Phenyl di-n-pentylphosphinate is a reasonably stable easily synthesized inhibitor of neuropathy target esterase (NTE) with low anticholinesterase activity. Like phenylmethylsulphonyl fluoride it protects hens against neuropathic effects of compounds such as diisopropylphosphorofluoridate. At intervals up to 15 days after dosing
Ntakadzeni E Madala et al.
Biotechnology letters, 34(7), 1351-1356 (2012-03-30)
Nicotiana tabacum cell suspensions, 2 g wet wt/ml, rapidly took up 1 mM isonitrosoacetophenone (INAP), a plant-derived stress metabolite with anti-oxidative and anti-fungal properties, producing 4'-hexopyranosyloxy-3'-methoxyisonitrosoacetophenone in 54 % yield over 18 h. Unconverted INAP was at 33 μM. UPLC-MS/MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service