Skip to Content
Merck
All Photos(1)

Key Documents

F3606

Sigma-Aldrich

3-Fluoroaniline

99%

Synonym(s):

1-Amino-3-fluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NH2
CAS Number:
Molecular Weight:
111.12
Beilstein:
1305471
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.542 (lit.)

bp

186 °C/756 mmHg (lit.)

density

1.156 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc(F)c1

InChI

1S/C6H6FN/c7-5-2-1-3-6(8)4-5/h1-4H,8H2

InChI key

QZVQQUVWFIZUBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N H Cnubben et al.
Chemico-biological interactions, 85(2-3), 151-172 (1992-12-01)
The in vitro and in vivo metabolism of monofluoroanilines was investigated. Special attention was focused on the regioselectivity of the aromatic hydroxylation by cytochromes P-450 and the mechanism by which this reaction might proceed. The results clearly demonstrate that the
The many roles for fluorine in medicinal chemistry.
William K Hagmann
Journal of medicinal chemistry, 51(15), 4359-4369 (2008-06-24)
Stéphanie Romand et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 63, 14-21 (2014-07-06)
A capillary isoelectric focusing (cIEF) method was developed for the determination of the ionization constants (pKa) of small molecules. Two approaches used to decrease the electroosmotic flow (EOF) were compared: (i) a hydroxypropylcellulose (HPC) coated capillary in aqueous medium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service