All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20
Beilstein:
79816
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
Assay
96%
refractive index
n20/D 1.451 (lit.)
bp
143 °C (lit.)
density
0.85 g/mL at 25 °C (lit.)
SMILES string
CCC1CCCCN1
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service