Skip to Content
Merck
All Photos(1)

Key Documents

DWKCTA0500P

Azlon Hexagonal Base PMP Cylinders with Printed Graduations

class B, capacity 500 mL

Synonym(s):

CTA0500P

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
€79.30

€79.30


Please contact Customer Service for Availability


Select a Size

Change View
2 EA
€79.30

About This Item

UNSPSC Code:
41121805
NACRES:
NA.54

€79.30


Please contact Customer Service for Availability

feature

class B
graduated

packaging

pkg of 2 ea

manufacturer/tradename

Azlon CTA0500P

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the measurement and dispensing of liquids in the laboratory

Features and Benefits

Calibrated to ISO 6706 and DIN 12681 class A tolerances

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service