Skip to Content
Merck
All Photos(1)

Key Documents

CLS6795620

Corning® hotplate stirrer, model PC-620

AC/DC input 120 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.72

stirring range measuring range

60-1100 rpm

temperature measuring range

77-1022 °F

L × W × D

4.8 in. × 10.5 in. × 14.4 in.

plate W × L

10 in. × 10 in.

Looking for similar products? Visit Product Comparison Guide

General description

The flat, bright-white Pyroceram® glass-ceramic top provides uniform heating and better contrast during applications that require monitoring of color changes. Pyroceram tops are easy to clean and highly resistant to scratches, corrosion, and chemicals. They can also be used at much higher temperatures (up to 550 °C) and are less likely to corrode than metal tops. The Pyroceram glass-ceramic material was originally developed by Corning for use in missile nosecones.

Legal Information

Corning is a registered trademark of Corning, Inc.
Pyroceram is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service