Documents
BR758902
BRAND® press-on lid for urine beaker
PE, red
Synonym(s):
Accessories, Lids, Press-on Lids, Sample Cups, Urine beaker
About This Item
UNSPSC Code:
41121814
NACRES:
NB.43
material
PE
red
manufacturer/tradename
BRAND 758902
packaging
pkg of (2 bags of 500)
General description
The BRAND® Press-On Lid for Urine Beakers is designed for clean and hygienic handling of urine samples, effectively preventing odors. Snap cap made of polyethylene (PE), this lid offers a secure closure that can be easily applied by simply pressing it on—no need for manual opening in the laboratory. For added convenience, the lid features a protrusion that can be easily cut to allow for the insertion of test strips or to pour out the contents.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG