Skip to Content
Merck
All Photos(1)

Documents

BMSD113328

Benchmark Stainless Steel Bulk Beads

bead diam. 2.8 mm, acid washed

Synonym(s):

Benchmark Stainless Steel Bulk Beads, grinding medium, grinding beads, beads for homogenization

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

stainless steel

packaging

pkg of 1,000 ea

manufacturer/tradename

Benchmark Scientific D1133-28

bead diam.

2.8 mm

compatibility

BeadBug and BeadBlaster

Looking for similar products? Visit Product Comparison Guide

General description

Loose, bulk beads, stainless steel,2.8mm diameter, acid washed, for bead homogenization

Application

bead, homogenization, lysis, stainless steel, TriplePure, BeadBug, BeadBlaster

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Repr. 1B - Skin Sens. 1 - STOT RE 2

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service