Skip to Content
Merck
All Photos(2)

Key Documents

94624

Sigma-Aldrich

S-(1-Oxido-2-pyridyl)-N,N,N′,N′-tetramethylthiuronium hexafluorophosphate

≥95.0% (NMR)

Synonym(s):

N,N,N′,N′-Tetramethyl-S-(1-oxido-2-pyridyl)thiuronium hexafluorophosphate, S-(2-Pyridyl)-N,N,N′,N′-tetramethylthiuronium N-oxide hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16F6N3OPS
CAS Number:
Molecular Weight:
371.28
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (NMR)

form

crystals

reaction suitability

reaction type: Coupling Reactions

mp

120-125 °C

application(s)

peptide synthesis

functional group

amine
thiourea

storage temp.

−20°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)\C(=[S+]\c1cccc[n+]1[O-])N(C)C

InChI

1S/C10H16N3OS.F6P/c1-11(2)10(12(3)4)15-9-7-5-6-8-13(9)14;1-7(2,3,4,5)6/h5-8H,1-4H3;/q+1;-1

InChI key

UCOGEMMJHLHOAW-UHFFFAOYSA-N

Application

Reagent for:
Synthesis of inhibitors for PAD1, 3, and 4
Synthesis of enantiomerically pure vinylcyclopropylboronic esters
Synthesis of enantiomerically pure cyclopropane building blocks
Camphor-derived chiral auxiliary for hydroxyalkyl radicals
Solution and solid-phase coupling
Conversion of carboxylic acids to Weinreb amides and N-methoxy or N-benzoxy amides

Other Notes

Very reactive peptide coupling reagent[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Mercaptopyridone 1-Oxide-Based Uronium Salts: New Peptide Coupling Reagents(1)(,)(2).
Miguel A. Bailén et al.
The Journal of organic chemistry, 64(24), 8936-8939 (2001-10-25)
F. Albericio et al.
Tetrahedron, 57, 9607-9607 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service