Skip to Content
Merck
All Photos(1)

Documents

922722

Sigma-Aldrich

O-Pivaloylhydroxyamine triflic acid

Synonym(s):

2,2-dimethyl-, azanyl ester, 1,1,1-trifluoromethanesulfonate, PivONH3OTF, Propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2 · CHF3O3S
CAS Number:
Molecular Weight:
267.22
UNSPSC Code:
12352106
NACRES:
NA.22

form

solid

Quality Level

Application

This hydroxylamine-based reagent enables the iron-catalyzed selective transformation of thiols to sulfinamides under mild conditions without the use of precious metal catalysts or additional oxidants. This reagent is shelf stable and facilitates the reaction in a single step and under mild conditions.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayanti Chatterjee et al.
Angewandte Chemie (International ed. in English), 60(2), 758-765 (2020-09-22)
An iron catalyzed reaction for the selective transformation of thiols (-SH) to sulfinamides (-SONH2 ) by a direct transfer of -O and free -NH2 groups has been developed. The reaction operates under mild conditions using a bench stable hydroxylamine derived
Eric Falk et al.
Organic letters, 23(4), 1422-1426 (2021-02-06)
We report both an intermolecular C-H amination of arenes to access N-methylanilines and an intramolecular variant for the synthesis of tetrahydroquinolines. A newly developed, highly electrophilic aminating reagent was key for the direct synthesis of unprotected N-methylanilines from simple arenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service