Skip to Content
Merck
All Photos(1)

Key Documents

860999

Sigma-Aldrich

Crystal Violet

certified by the Biological Stain Commission, Dye content 88 %

Synonym(s):

Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
Colour Index Number:
42555
Beilstein:
3580948
EC Number:
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

grade

certified by the Biological Stain Commission

composition

Dye content, 88%

mp

205 °C (dec.) (lit.)

λmax

588 nm

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

A modified Gram stain for differentiating between Gram-positive and Gram-negative organisms, using Safranine O as counterstain.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service