Skip to Content
Merck
All Photos(1)

Documents

802964

Sigma-Aldrich

1-Chloro-3,5-dimethyladamantane

95%

Synonym(s):

1,3-Dimethyl-5-chloroadamantane, 1-chloro-3,5-dimethyl-Tricyclo[3.3.1.13,7]decane, 5-Chloro-1,3-dimethyladamantane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19Cl
CAS Number:
Molecular Weight:
198.73
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

Flash Point: 215°F

Quality Level

Assay

95%

form

liquid

refractive index

n/D 1.4962

density

1.0175 g/mL at 25 °C

SMILES string

C[C@]12CC3C[C@](C)(C1)CC(Cl)(C3)C2

InChI

1S/C12H19Cl/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8H2,1-2H3/t9-,10+,11-,12-

InChI key

PXDRFQZLDWZHPX-CDECOKDKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Building block has been shown to be used as a key intermediate in the synthesis of memantine hydrochloride, which was reported to be a FDA approved drug for Alzheimer′s treatment.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F

Flash Point(C)

102 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service