Skip to Content
Merck
All Photos(1)

Key Documents

797464

Sigma-Aldrich

7-Amino-5-oxa-2-azaspiro[3.4]octane-2-carboxylic acid 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20N2O3
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

color

faint yellow

functional group

ether

storage temp.

2-8°C

SMILES string

NC(CO1)CC21CN(C(OC(C)(C)C)=O)C2

InChI

1S/C11H20N2O3/c1-10(2,3)16-9(14)13-6-11(7-13)4-8(12)5-15-11/h8H,4-7,12H2,1-3H3

InChI key

DJQMEIQMZYMIIH-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers, which provides a new area of chemical space with straightforward functional handles for further diversification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service