Skip to Content
Merck
All Photos(1)

Key Documents

777714

Sigma-Aldrich

3-Amino-4-bromo-1H-pyrazole

95%

Synonym(s):

4-Bromo-1H-pyrazol-3-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4BrN3
CAS Number:
Molecular Weight:
161.99
UNSPSC Code:
12352100
NACRES:
NA.22
form:
solid
Assay:
95%

Assay

95%

form

solid

mp

135-140 °C

InChI

1S/C3H4BrN3/c4-2-1-6-7-3(2)5/h1H,(H3,5,6,7)

InChI key

OELYMZVJDKSMOJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service