Skip to Content
Merck
All Photos(1)

Key Documents

773492

Sigma-Aldrich

Poly(N-[2-(propylamino)ethyl]-10,12-pentacosadiyneamide hydrogen bromide) nanotube

>99% (SEM)

Synonym(s):

PDNT, PDNT-12-8-23Br, Polydiacetylene Nanotube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

Assay

>99% (SEM)

form

powder

O.D. × I.D. × L

110 nm × 40 nm × 1,000 nm

General description

Alignment of the self assembled diacetylene monomers render well polymerized polydiacetylene nanotubes (PDNTs). SEM images reveal contamination free nanotube lumen openings.

Application

  • Stimuli-sensitive smart nanomaterial composites
  • Drug delivery systems(DDS)
  • Biosensors for viruses; bacteria; andglucose
  • Langmuir-Blodgett films
  • Lab-on-a-chip
  • Colorimetric pressure sensors
  • UV-based sensors
  • Nanoelectronics
  • Optics
  • Artificial retinas
  • Neuronal networks
  • MEMS
  • Membrane filtration
  • Gas separation

Analysis Note

Appearance: Solid deep blue powder at room temperature
Purity: >99% (SEM)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polydiacetylene paper-based colorimetric sensor array for vapor phase detection and identification of volatile organic compounds
Journal of Materials Chemistry, 22, 5970-5970 (2012)
Sang Beom Lee et al.
Journal of the American Chemical Society, 126(41), 13400-13405 (2004-10-14)
We describe the facile two-step synthesis of nanotubes that form pure, well-defined, nanostructured materials. We have synthesized a secondary amine HBr salt as the headgroup of a single-chain diacetylenic lipid. This molecule can form a number of different self-assembled nanostructures
Drug Delivery and Imaging with Polydiacetylene Micelles
Chemistry (Weinheim An Der Bergstrasse, Germany), 18, 400-408 (2012)
Sang Beom Lee et al.
Nano letters, 5(11), 2202-2206 (2005-11-10)
We describe here the first method for dispersion of individual self-assembled diacetylene nanotubes on surfaces. Complete polymerization by UV exposure was achieved as demonstrated by nanotubes that were resistant to aggressive organic solvents and temperatures well above the melting point

Articles

Polydiacetylene nanotubes (PDNTs) are self-assembled diacetylene nanotubes comprised of cross-linked, conjugated double and triple bonds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service