767166
Polyester bis-MPA dendron, 16 carboxyl, 1 amine
generation 4
Synonym(s):
bisMPA dendrimer, bisMPA dendron
About This Item
Recommended Products
description
carboxyl surface groups
form
solid
mol wt
generation 4
feature
focal point amine
no. Surface Groups
16
storage temp.
−20°C
InChI
1S/C147H195NO94/c1-16-147(81-228-118(197)84-17-19-85(148)20-18-84,82-241-131(210)145(14,77-237-127(206)141(10,69-229-119(198)133(2,53-212-102(181)37-21-86(149)150)54-213-103(182)38-22-87(151)152)70-230-120(199)134(3,55-214-104(183)39-23-88(153)154)56-215-105(184)40-24-89(155)156)78-238-128(207)142(11,71-231-121(200)135(4,57-216-106(185)41-25-90(157)158)58-217-107(186)42-26-91(159)160)72-232-122(201)136(5,59-218-108(187)43-27-92(161)162)60-219-109(188)44-28-93(163)164)83-242-132(211)146(15,79-239-129(208)143(12,73-233-123(202)137(6,61-220-110(189)45-29-94(165)166)62-221-111(190)46-30-95(167)168)74-234-124(203)138(7,63-222-112(191)47-31-96(169)170)64-223-113(192)48-32-97(171)172)80-240-130(209)144(13,75-235-125(204)139(8,65-224-114(193)49-33-98(173)174)66-225-115(194)50-34-99(175)176)76-236-126(205)140(9,67-226-116(195)51-35-100(177)178)68-227-117(196)52-36-101(179)180/h17-20H,16,21-83,148H2,1-15H3,(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H,159,160)(H,161,162)(H,163,164)(H,165,166)(H,167,168)(H,169,170)(H,171,172)(H,173,174)(H,175,176)(H,177,178)(H,179,180)
InChI key
JTLTWQVZAUYNJV-UHFFFAOYSA-N
General description
Application
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service