Skip to Content
Merck
All Photos(1)

Key Documents

745928

Sigma-Aldrich

PDTSTPD

Synonym(s):

Poly[(5,6-dihydro-5-octyl-4,6-dioxo-4H-thieno[3,4-c]pyrrole-1,3-diyl)[4,4-bis(2-ethylhexyl)-4H-silolo[3,2-b:4,5-b′;]dithiophene-2,6-diyl]]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C38H53NO2S3Si)n
CAS Number:
UNSPSC Code:
12352103

description

Band gap: 1.8 eV

form

powder

mol wt

average Mn 7,000-35,000 by GPC

λmax

670 nm (thin film)

Orbital energy

HOMO -5.6 eV 
LUMO -3.8 eV 

semiconductor properties

P-type (mobility=1.0 x 10-4 cm2/V·s) (OFET)

General description

HOMO: -5.6 eV
LUMO: -3.8 eV
UV absorption: λmax = 670 nm; Eg = 1.73 eV
Solubility: This polymer can be dissolved easily in chloroform, chlorobenzene, dichlorobenzene

Application

High-Efficiency Organic Solar Cells (OPVs)
Power conversion efficiency (PCE): 7.3%
Open circuit voltage (Voc): 0.88 V
Short circuit current density (Jsc): 12.2 mA/cm2
Fill factor (FF): 0.69

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ta-Ya Chu et al.
Journal of the American Chemical Society, 133(12), 4250-4253 (2011-03-08)
A new alternating copolymer of dithienosilole and thienopyrrole-4,6-dione (PDTSTPD) possesses both a low optical bandgap (1.73 eV) and a deep highest occupied molecular orbital energy level (5.57 eV). The introduction of branched alkyl chains to the dithienosilole unit was found
Xugang Guo et al.
Journal of the American Chemical Society, 134(44), 18427-18439 (2012-10-04)
Rational creation of polymeric semiconductors from novel building blocks is critical to polymer solar cell (PSC) development. We report a new series of bithiopheneimide-based donor-acceptor copolymers for bulk-heterojunction (BHJ) PSCs. The bithiopheneimide electron-deficiency compresses polymer bandgaps and lowers the HOMOs--essential

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service