Skip to Content
Merck
All Photos(1)

Documents

732737

Sigma-Aldrich

4-(Benzaldehyde diethylacetal)magnesium bromide solution

1 M in THF

Synonym(s):

4-(Diethoxymethyl)phenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BrMgO2
Molecular Weight:
283.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: Grignard Reaction

concentration

1 M in THF

density

1.013 g/mL at 25 °C

SMILES string

CCOC(OCC)c1ccc([Mg]Br)cc1

InChI

1S/C11H15O2.BrH.Mg/c1-3-12-11(13-4-2)10-8-6-5-7-9-10;;/h6-9,11H,3-4H2,1-2H3;1H;/q;;+1/p-1

InChI key

APPJPCOJOCVSMA-UHFFFAOYSA-M

Application

4-(Benzaldehyde diethylacetal)magnesium bromide can be used as a substrate in the sp3 C-H arylation of tetrahydroisoquinolines using 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

-30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple and Direct sp3 C-H Bond Arylation of Tetrahydroisoquinolines and Isochromans via 2, 3-Dichloro-5, 6-dicyano-1, 4-benzoquinone Oxidation under Mild Conditions
Muramatsu W, et al.
Organic Letters, 15(14), 3650-3653 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service