Skip to Content
Merck
All Photos(1)

Key Documents

72801

Sigma-Aldrich

4-Nitrobenzaldehyde oxime

≥97.0% (N)

Synonym(s):

4-Nitrobenzaldoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH=NOH
CAS Number:
Molecular Weight:
166.13
Beilstein:
2047086
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (N)

mp

126-130 °C (lit.)
128-131 °C

SMILES string

[H]\C(=N\O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H6N2O3/c10-8-5-6-1-3-7(4-2-6)9(11)12/h1-5,10H/b8-5+

InChI key

WTLPAVBACRIHHC-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent (as tetramethylguanidine salt) for the cleavage of the 2-chlorophenyl protection in oligonucleotides in high yield with little or no nucleotide cleavage

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.B. Reese et al.
Tetrahedron Letters, 2727-2727 (1978)
Tetrahedron Letters, 4443-4443 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service