Skip to Content
Merck
All Photos(1)

Documents

709247

Sigma-Aldrich

Zinc arsenide

pieces, 99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn3As2
CAS Number:
Molecular Weight:
346.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

pieces

mp

1015 °C

density

5.53 g/mL at 25 °C (lit.)

SMILES string

[Zn]=[As][Zn][As]=[Zn]

InChI

1S/2As.3Zn

InChI key

RHKSESDHCKYTHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service