Skip to Content
Merck
All Photos(1)

Key Documents

708844

Sigma-Aldrich

(1S,2S)-N,N′-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)

Synonym(s):

(S,S)-AMAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H38CoN2O4
CAS Number:
Molecular Weight:
573.59
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

optical activity

[α]/D −309°, c = 0.1 in chloroform

mp

242-246 °C

functional group

ketone

SMILES string

CC(=O)C1=C(C)O[Co]OC(C)=C(\C=N\[C@H]([C@@H](\N=C\1)c2c(C)cc(C)cc2C)c3c(C)cc(C)cc3C)C(C)=O

InChI

1S/C32H40N2O4.Co/c1-17-11-19(3)29(20(4)12-17)31(33-15-27(23(7)35)24(8)36)32(34-16-28(25(9)37)26(10)38)30-21(5)13-18(2)14-22(30)6;/h11-16,31-32,35,37H,1-10H3;/q;+2/p-2/b27-23+,28-25+,33-15+,34-16+;/t31-,32-;/m0./s1

InChI key

PHCQQLMRNZRDJA-OZXAGROVSA-L

Application

Catalyst for:
  • Stereoselective preparation of benzylic alcohols via oxidative kinetic resolution of secondary benzylic alcohols with oxygen in presence of alkene
  • Enantioselective borohydride reduction
  • Preparation of ß-nitro alcohols from aldehydes and nitroalkanes by stereoselective Henry reaction in presence of amine bases
  • Preparation of optically pure diarylpropanediols via reduction of the corresponding diketones

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service