Skip to Content
Merck
All Photos(1)

Documents

694010

Sigma-Aldrich

1H-Benzo[g]indole-3-carboxaldehyde

97%

Synonym(s):

3-Formyl-1H-benzo[g]indole, 3-Formyl-6,7-benzindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

231-236 °C

SMILES string

O=Cc1c[nH]c2c1ccc3ccccc23

InChI

1S/C13H9NO/c15-8-10-7-14-13-11-4-2-1-3-9(11)5-6-12(10)13/h1-8,14H

InChI key

NYIGHGXJJVOYCQ-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
Useful starter in indole and natural product synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 2450-2450 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service