Skip to Content
Merck
All Photos(3)

Key Documents

681636

Sigma-Aldrich

4-Fluoro-1-indanone

97%

Synonym(s):

4-Fluoro-2,3-dihydro-1-indenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7FO
CAS Number:
Molecular Weight:
150.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

72-76 °C

functional group

fluoro
ketone

SMILES string

Fc1cccc2C(=O)CCc12

InChI

1S/C9H7FO/c10-8-3-1-2-7-6(8)4-5-9(7)11/h1-3H,4-5H2

InChI key

HOMSJDBZHCPYHY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service