Skip to Content
Merck
All Photos(1)

Documents

673390

Sigma-Aldrich

1,3-Di-tert-butylbenzimidazolium chloride

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23ClN2
CAS Number:
Molecular Weight:
266.81
MDL number:
UNSPSC Code:
12352005
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

reaction suitability

reagent type: catalyst

mp

204 °C (dec.)

SMILES string

[Cl-].CC(C)(C)n1c[n+](c2ccccc12)C(C)(C)C

InChI

1S/C15H23N2.ClH/c1-14(2,3)16-11-17(15(4,5)6)13-10-8-7-9-12(13)16;/h7-11H,1-6H3;1H/q+1;/p-1

InChI key

CMQDFXSMZLANNA-UHFFFAOYSA-M

Application

Reactant for preparation of:
  • NHC borane complexes by Lewis base exchange
  • Fluorescent 5,5′-bibenzimidazolylidenes
  • Donor-acceptor triazenes

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Emerging class of privileged ligands

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service