663786
N,N-Diethyl-1H-indole-1-carboxamide
97%
Synonym(s):
Indole-1-carboxylic acid diethylamide
About This Item
Assay
97%
refractive index
n20/D 1.575
bp
140-147 °C/0.8 mmHg
density
1.089 g/mL at 25 °C
SMILES string
CCN(CC)C(=O)n1ccc2ccccc12
InChI
1S/C13H16N2O/c1-3-14(4-2)13(16)15-10-9-11-7-5-6-8-12(11)15/h5-10H,3-4H2,1-2H3
InChI key
DNEILLNCDATSFX-UHFFFAOYSA-N
Application
- reactant in preparation of substituted indoles via ortho-metalation and Suzuki-Miyaura cross coupling
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service