Skip to Content
Merck
All Photos(1)

Key Documents

659649

Sigma-Aldrich

Polylactide-block-poly(ethylene glycol)-block-polylactide

PLA average Mn 1,000, PEG average Mn 10,000

Synonym(s):

PLA-PEG-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CHCH3COO)x(CH2CH2O)y(COCHCH3O)zH
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 10,000
PLA average Mn 1,000
average Mn (1,000-10,000-1,000)

degradation timeframe

<12 months

PDI

<1.2

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Biodegradable polymer. Applications include drug encapsulation and drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service