All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H14ClFN2
CAS Number:
Molecular Weight:
228.69
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.5470 (lit.)
density
1.202 g/mL at 25 °C (lit.)
SMILES string
Fc1cccc(Cl)c1CN2CCNCC2
InChI
1S/C11H14ClFN2/c12-10-2-1-3-11(13)9(10)8-15-6-4-14-5-7-15/h1-3,14H,4-8H2
InChI key
PBJVZHBDDDMHIT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service