Skip to Content
Merck
All Photos(1)

Key Documents

647683

Sigma-Aldrich

2-Amino-5-bromobenzothiazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrN2S
CAS Number:
Molecular Weight:
229.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
form:
solid
Assay:
96%
Pricing and availability is not currently available.

Assay

96%

form

solid

mp

194-198 °C

functional group

bromo

SMILES string

Nc1nc2cc(Br)ccc2s1

InChI

1S/C7H5BrN2S/c8-4-1-2-6-5(3-4)10-7(9)11-6/h1-3H,(H2,9,10)

InChI key

ZPUJTWBWSOOMRP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire Bagnéris et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8428-8433 (2014-05-23)
Voltage-gated sodium channels are important targets for the development of pharmaceutical drugs, because mutations in different human sodium channel isoforms have causal relationships with a range of neurological and cardiovascular diseases. In this study, functional electrophysiological studies show that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service