Skip to Content
Merck
All Photos(1)

Documents

614785

Sigma-Aldrich

Benzyl-α,α-d2 chloride

98 atom % D

Synonym(s):

α-Chlorotoluene-α,α−d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CD2Cl
Molecular Weight:
128.60
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

bp

177-181 °C (lit.)

mp

-43 °C (lit.)

density

1.117 g/mL at 25 °C

mass shift

M+2

SMILES string

ClC([2H])([2H])C1=CC=CC=C1

InChI

1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2/i6D2

InChI key

KCXMKQUNVWSEMD-NCYHJHSESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Heart,forestomach, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service