Skip to Content
Merck
All Photos(1)

Key Documents

588962

Sigma-Aldrich

4,6-Dichloro-1,3,5-triazine on silica gel

200-400 mesh, extent of labeling: 0.5-1.0 mmol/g loading

Synonym(s):

Cyanuric acid on silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

extent of labeling

0.5-1.0 mmol/g loading

particle size

200-400 mesh

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service