Skip to Content
Merck
All Photos(1)

Key Documents

553417

Sigma-Aldrich

Aluminum chloride THF complex

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8AlCl3O
CAS Number:
Molecular Weight:
205.45
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
liquid

grade

for analytical purposes

form

liquid

reaction suitability

reagent type: catalyst
core: aluminum

concentration

0.5 M in THF

bp

65 °C (lit.)

density

0.953 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Cl[Al](Cl)Cl.C1CCOC1

InChI

1S/C4H8O.Al.3ClH/c1-2-4-5-3-1;;;;/h1-4H2;;3*1H/q;+3;;;/p-3

InChI key

QBBWRNBMWSVXBB-UHFFFAOYSA-K

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service