Skip to Content
Merck
All Photos(1)

Key Documents

546909

Sigma-Aldrich

2-Bromo-5-fluorobenzotrifluoride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CF3
CAS Number:
Molecular Weight:
243.00
Beilstein:
2643544
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.465 (lit.)

bp

136-143 °C (lit.)

density

1.695 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1ccc(Br)c(c1)C(F)(F)F

InChI

1S/C7H3BrF4/c8-6-2-1-4(9)3-5(6)7(10,11)12/h1-3H

InChI key

AIDVAZGOACECLJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.0 °F - closed cup

Flash Point(C)

63.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service