Skip to Content
Merck
All Photos(2)

Key Documents

542245

Sigma-Aldrich

3′,5′-Dibromo-4′-hydroxyacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H2(OH)COCH3
CAS Number:
Molecular Weight:
293.94
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

182-187 °C (lit.)

SMILES string

CC(=O)c1cc(Br)c(O)c(Br)c1

InChI

1S/C8H6Br2O2/c1-4(11)5-2-6(9)8(12)7(10)3-5/h2-3,12H,1H3

InChI key

ZNWPTJSBHHIXLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3′,5′-Dibromo-4′-hydroxyacetophenone (3,5-dibromo-4-hydroxyacetophenone) can be obtained from 4-hydroxyacetophenone, via bromination by N-bromosuccinimide in the presence of LiClO4-SiO2.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An intriguing effect of lithium perchlorate dispersed on silica gel in the bromination of aromatic compounds by N-bromosuccinimide.
Bagheri M, et al.
Canadian Journal of Chemistry, 83(2), 146-149 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service