Skip to Content
Merck
All Photos(1)

Key Documents

541826

Sigma-Aldrich

Bis(3,5-dichlorophenyl) disulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cl2C6H3S)2
CAS Number:
Molecular Weight:
356.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

66-69 °C (lit.)

SMILES string

Clc1cc(Cl)cc(SSc2cc(Cl)cc(Cl)c2)c1

InChI

1S/C12H6Cl4S2/c13-7-1-8(14)4-11(3-7)17-18-12-5-9(15)2-10(16)6-12/h1-6H

InChI key

JMQANWHMOHXBEA-UHFFFAOYSA-N

General description

Bis(3,5-dichlorophenyl) disulfide is also known as 3,3′,5,5′-tetrachlorodiphenyl disulfide.

Application

Bis(3,5-dichlorophenyl) disulphide may be used in the synthesis of 3-(3-((3,5-dichlorophenyl)thio)-6-methyl-1H-indol-2-yl)propanoic acid.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manunya Nuth et al.
Journal of medicinal chemistry, 56(8), 3235-3246 (2013-03-27)
Smallpox constitutes a major bioterrorism threat, which underscores the need to develop antiviral drugs for rapid response in the event of an attack. Viral processivity factors are attractive drug targets in being both specific and essential for their cognate DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service