Skip to Content
Merck
All Photos(1)

Key Documents

47318

Sigma-Aldrich

p-Fluoro-D-phenylalanine

≥99.0% (sum of enantiomers, HPLC)

Synonym(s):

4-Fluoro-D-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10FNO2
CAS Number:
Molecular Weight:
183.18
Beilstein:
2416149
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (sum of enantiomers, HPLC)

form

solid

optical activity

[α]20/D +23±2°, c = 1% in H2O

optical purity

enantiomeric ratio: ≥99:1 (TLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

~245 °C (dec.)

application(s)

peptide synthesis

SMILES string

N[C@H](Cc1ccc(F)cc1)C(O)=O

InChI

1S/C9H10FNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m1/s1

InChI key

XWHHYOYVRVGJJY-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service