Skip to Content
Merck
All Photos(2)

Documents

457655

Sigma-Aldrich

Poly(dimethylsiloxane-co-alkylmethylsiloxane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Si(CH3)2O]x[Si(CH3)(R)O]y, R = C16-C18
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

20 °C (lit.)

transition temp

softening point >23 °C

solubility

hydrocarbons, higher alcohols and fatty esters: soluble

density

0.85 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(O)O.CCCCCCCCCCCCCCCC[Si](C)(O)O

Features and Benefits

Good lubricity and spreadability,

Physical form

The copolymer is linear polysiloxane functionalized with (~32) branched and linear alkyl groups.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service