Skip to Content
Merck
All Photos(1)

Key Documents

447781

Sigma-Aldrich

5-(3-Nitrophenyl)-2-furoic acid

95%

Synonym(s):

5-(3-Nitrophenyl)-2-furancarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7NO5
CAS Number:
Molecular Weight:
233.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

252-260 °C (lit.)

functional group

carboxylic acid
nitro

SMILES string

OC(=O)c1ccc(o1)-c2cccc(c2)[N+]([O-])=O

InChI

1S/C11H7NO5/c13-11(14)10-5-4-9(17-10)7-2-1-3-8(6-7)12(15)16/h1-6H,(H,13,14)

InChI key

XWQLGLUTQIKSAJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyoung Gee Baek et al.
Journal of combinatorial chemistry, 11(1), 91-102 (2008-12-09)
The one-bead one-compound (OBOC) combinatorial library method enables the rapid generation and screening of millions of discrete chemical compounds on beads. Most of the OBOC screening methods require the library compounds to remain tethered to the bead during screening process.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service