Skip to Content
Merck
All Photos(1)

Documents

428973

Sigma-Aldrich

1-Hepten-3-ol

97%

Synonym(s):

Butyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH=CH2
CAS Number:
Molecular Weight:
114.19
Beilstein:
1720340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.433 (lit.)

bp

153-156 °C (lit.)

density

0.836 g/mL at 20 °C
0.836 g/mL at 25 °C (lit.)

SMILES string

CCCCC(O)C=C

InChI

1S/C7H14O/c1-3-5-6-7(8)4-2/h4,7-8H,2-3,5-6H2,1H3

InChI key

PZKFYTOLVRCMOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hepten-3-ol (HEPT) is an allylic alcohol. It is one of the volatile compounds found in mentha citrate, flowers of Hyacinthus orientalis L. and green table olive fruits. It is reported to be a potent odor stimulus present in the human volatile emanations. 1-Hepten-3-ol is reported to elicit behavioral responses in Anopheles gambiae. Its mass spectra has been analyzed.

Application

1-Hepten-3-ol (HEPT) may be used in the synthesis of 1,1,1-trichloro-4-octanone.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The formation of [CH3CH2C(=OH)CH2]+. from 1-hepten-3-ol.
McAdoo DJ and Hudson CE.
Org. Mass Spectrom., 16(7), 294-296 (1981)
Yu Tong Qiu et al.
Chemical senses, 31(9), 845-863 (2006-09-12)
Olfactory receptor neurons (ORNs) in the antenna of insects serve to encode odors in action potential activity conducted to the olfactory lobe of the deuterocerebrum. We performed an analysis of the electrophysiological responses of olfactory neurons in the antennae of
A Palladium Catalyzed Conversion of Halohydrins to Ketones.
Jiro T, et al.
Tetrahedron Letters, 23(30), 3085-3088 (1982)
Free and glycosidically bound volatiles of Mentha citrata ehrh.
Mastelic J, et al.
Croatica Chemica Acta. Arhiv Za Kemiju, 73(3), 781-794 (2000)
K Hosokawa et al.
Plant cell reports, 14(9), 575-579 (1995-06-01)
Essential oils were produced by flowers of Hyacinthus orientalis L. that had been regenerated in vitro. The production of these oils was affected by the concentration of gibberellic acid and sucrose in the medium and by temperature. The highest concentration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service