425494
Boron-10B trifluoride diethyl etherate
99 atom % 10B
About This Item
isotopic purity
99 atom % 10B
refractive index
n20/D 1.344 (lit.)
bp
126 °C (lit.)
density
1.114 g/mL at 25 °C (lit.)
mass shift
M-1
SMILES string
F[10B](F)F.CCOCC
InChI
1S/C4H10O.BF3/c1-3-5-4-2;2-1(3)4/h3-4H2,1-2H3;/i;1-1
InChI key
KZMGYPLQYOPHEL-HCMAANCNSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT RE 1 Inhalation
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
118.4 °F - closed cup
Flash Point(C)
48.00 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service