Skip to Content
Merck
All Photos(1)

Key Documents

421413

Sigma-Aldrich

Zonyl® FSN fluorosurfactant

Synonym(s):

nonionic fluorosurfactant

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)x(CF2)yC2H5FO
CAS Number:
MDL number:
UNSPSC Code:
12162002

refractive index

n20/D 1.3820

bp

80 °C

density

1.06 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service