Skip to Content
Merck
All Photos(1)

Key Documents

40289

Sigma-Aldrich

Acetone

semiconductor grade VLSI PURANAL (Honeywell 17617)

Synonym(s):

Dimethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021201
PubChem Substance ID:
grade:
semiconductor grade VLSI PURANAL (Honeywell 17617)
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

semiconductor grade VLSI PURANAL (Honeywell 17617)

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

form

liquid

CofA

specification on request

expl. lim.

13.2 %

color

clear colorless

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
Semiconductor grade acetone may be used for cleaning of the surfaces, such as;
  • Hydrogen terminated silicon wafer.[1][2]
  • SiC wafers.[3]
  • Chromium nitride epilayer on silicon wafer.[4]

The semiconductor grade acetone was also used in a comparative study for observing the effect of various solvents on nitrogen containing promoters (NCPs), which is used in oxidation reaction of 5-hydroxymethylfurfural.[5]

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalently attached organic monolayers onto silicon carbide from 1-alkynes: molecular structure and tribological properties.
Pujari S, et al.
Langmuir, 29(13), 4019-4031 (2013)
Mono-fluorinated alkyne-derived SAMs on oxide-free Si (111) surfaces: preparation, characterization and tuning of the Si workfunction
Pujari S, et al.
Langmuir, 29(2), 570-580 (2013)
Cu catalyzed oxidation of 5-hydroxymethylfurfural to 2, 5-diformylfuran and 2, 5-furandicarboxylic acid under benign reaction conditions.
Hansen T, et al.
Applied Catalysis A: General, 456, 44-50 (2013)
Hexadecadienyl monolayers on hydrogen-terminated Si (111): faster monolayer formation and improved surface coverage using the enyne moiety
Rijksen B, et al.
Langmuir, 28(16), 6577-6588 (2012)
Organic monolayers from 1-alkynes covalently attached to chromium nitride: alkyl and fluoroalkyl termination.
Pujari S, et al.
Langmuir, 29(33), 10393-10404 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service